2-Methylbutyl acetate

Supplier: Spectrum Chemical Mfg. Corp.
Danger

M2578-500ML M2578-100ML M2578-25ML
700004-574EA 709.55 USD
700004-574 700004-570 700004-572
2-Methylbutyl acetate
2-Methylbutyl acetate
Formula: C₇H₁₄O₂
Boiling Pt: 140…142 °C (1013 hPa)
Density: 0.876 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00040494
CAS Number: 624-41-9
EINECS: 210-843-8
UN: 1104
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR