3-Methyl-1,2-cyclopentanedione ≥99%
Supplier: SPECTRUM CHEMICAL MFG CORP
M2243-10GM
M2243-50GM
700004-000EA
107.63
USD
700004-000
700004-002
3-Methyl-1,2-cyclopentanedione ≥99%
3-Methyl-1,2-cyclopentanedione
3-Methylcyclopentane-1,2-dione is a fragrance or odor agent, used in home products (cleaners, laundry products, air fresheners) or similar industrial products.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
C₆H₈O₂ Boiling Pt: 210 °C (1013 hPa) Storage Temperature: Ambient |
MDL Number:
MFCD00001417 CAS Number: 765-70-8 EINECS: 212-154-8 |
Specification Test Results
Assay | 99% Min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...