Magnesium silicate suitable for use as excipient

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Florisil

M1343-2.5KG M1343-12KG
700001-884EA 310.7 USD
700001-884 700001-886 700001-882
Magnesium silicate suitable for use as excipient
Magnesium silicate

Magnesium Silicate, also known as talc, is used as a lubricant in cosmetic preparations, a glidant in pharmaceutical tablets and as a filler in other applications.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: MgO₃Si
Storage Temperature: Ambient
MDL Number: MFCD00078249
CAS Number: 1343-88-0
EINECS: 215-681-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR