Methyl nicotinate ≥99%, Reagent Grade
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
3-Picolinic acid methyl ester, 3-(Methoxycarbonyl)pyridine, Pyridine-3-carboxylic acid methyl ester, Nicotinic acid methyl ester, 3-Pyridinecarboxylic acid methyl ester, Methyl 3-pyridinecarboxylate
M1276-100GM
M1276-500GM
M1276-12KGBL
M1276-5KGBL
700009-162EA
112
USD
700009-162
700009-166
700009-164
700009-168
Methyl nicotinate ≥99%, Reagent Grade
Methyl nicotinate
BSE/TSE statement (available upon request)
Allergen statement (available upon request)
Formula:
C₇H₇NO₂ MW: 137.14 g/mol Boiling Pt: 209 °C (1013 hPa) Melting Pt: 39 °C Density: 1.0048 g/cm³ (20 °C) Flash Pt: 95 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006388 CAS Number: 93-60-7 EINECS: 202-261-8 Merck Index: 12,06174 |
Specification Test Results
Assay | 99% Min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...