Benzoic acid methyl ester ≥98% for synthesis

Supplier: SPECTRUM CHEMICAL MFG CORP
Warning

Synonyms: Methyl benzoate

M1243-200LTB M1243-500MLG M1243-20LT
700005-588EA 5861.15 USD
700005-588 700005-594 700005-590
Benzoic acid methyl ester ≥98% for synthesis
Benzoic acid methyl ester

Methyl Benzoate, an organic compound and ester, can be used in perfumery, as a solvent, and as a solvent.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₆H₅COOCH₃
Boiling Pt: 199.6 °C (1013 hPa)
Melting Pt: –12 °C
Density: 1.0888 g/cm³ (20 °C)
Flash Pt: 83 °C
Storage Temperature: Ambient
MDL Number: MFCD00008421
CAS Number: 93-58-3
EINECS: 202-259-7
Merck Index: 13,06052
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=287e16081e44f5525ff8b4d34f0a6fece44a3073&vid=2be9af3b90e55af477c374c091cd0888dcf60b96&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 98% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR