Methyl acetate ≥96%, Reagent Grade

Supplier: SPECTRUM CHEMICAL MFG CORP
Danger

Synonyms: Acetic acid methyl ester

M1225-200LTB M1225-4LT M1225-500MLG M1225-1LT M1225-20LTBL
700004-538EA 2550.7 USD
700004-538 700004-542 700004-544 700004-536 700004-540
Methyl acetate ≥96%, Reagent Grade
Methyl acetate

Methyl Acetate, Reagent is also known as acetic acid methyl ester. It is primarily used as a volatile low toxicity solvent in the laboratory, paints, glues and nail polish removers. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.

Formula: C₃H₆O₂
MW: 74.08 g/mol
Boiling Pt: 56.9 °C (1013 hPa)
Melting Pt: –98 °C
Density: 0.938 g/cm³ (20 °C)
Flash Pt: –13 °C
Storage Temperature: Ambient
MDL Number: MFCD00008711
CAS Number: 79-20-9
EINECS: 201-185-2
UN: 1231
ADR: 3,II
Merck Index: 13,06038

Order Now

Specification Test Results

Assay 96% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR