Diethylene glycol monomethyl ether ≥98%, Reagent Grade for synthesis

Supplier: SPECTRUM CHEMICAL MFG CORP
Warning

Synonyms: 2-(2-Methoxyethoxy)ethanol, Diethylene glycol methyl ether

M1222-20LTMT M1222-4LT
700003-476EA 1210.25 USD
700003-476 700003-478
Diethylene glycol monomethyl ether ≥98%, Reagent Grade for synthesis
Diethylene glycol monomethyl ether

2-(2-Methoxyethoxy)ethanol, Reagent, also known as methyl carbitol, is used as an industrial solvent and can be used as a fuel system icing inhibitor in jet fuels. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.

Formula: C₅H₁₂O₃
MW: 120.15 g/mol
Boiling Pt: 193
Melting Pt: -65
Density: 1.010
Flash Pt: 91 °C
Storage Temperature: Ambient
MDL Number: MFCD00002871
CAS Number: 111-77-3
EINECS: 203-906-6
Merck Index: 12,06116

Order Now

Specification Test Results

Assay 98% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR