Maltol ≥99% FCC
Supplier: SPECTRUM CHEMICAL MFG CORP
|
Warning
|
Synonyms:
3-Hydroxy-2-methyl-γ-pyrone, Maltol, 3-Hydroxy-2-methyl-4-pyranone, 3-Hydroxy-2-methyl-4H-pyran-4-one, 3-Hydroxy-2-methyl-4-pyrone
Maltol, FCC is used as a flavor enhancer in breads and cakes. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formula:
C₆H₆O₃ MW: 126.11 g/mol Boiling Pt: 285 °C (1013 hPa) Density: 1.453 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00006578 CAS Number: 118-71-8 EINECS: 204-271-8 Merck Index: 13,05735 |
Specification Test Results
Assay | 99% Min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...