Magnesium oxide heavy ≥96%, powder FCC

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Magnesia usta

M1168-2.5KG M1168-500GM
95034-234EA 738.1 USD
95034-234 95034-236
Magnesium oxide heavy ≥96%, powder FCC
Magnesium oxide

Heavy Magnesium Oxide, Heavy, Powder, FCC is a natural, GRAS ingredient used as an anti-caking agent, firming agent and for nutrition fortification. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: MgO
MW: 40.3 g/mol
Boiling Pt: 3600 °C (1013 hPa)
Melting Pt: 2800 °C
Density: 3.58 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011109
CAS Number: 1309-48-4
EINECS: 215-171-9
Merck Index: 12,05713

Order Now

Specification Test Results

Assay 96% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR