Manganese(II) carbonate, powder, Reagent Grade
Supplier: Spectrum Chemical Mfg. Corp.
Manganese Carbonate, Powder, Reagent is widely used as an additive to plant fertilizers to cure manganese deficient crops and in health foods, in ceramics as a glaze colorant and flux, and in concrete stains. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.
Formula:
CMnO₃ Density: 3.12 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011116 CAS Number: 598-62-9 EINECS: 209-942-9 Merck Index: 13,05749 |
Specification Test Results
Assay | 43.5% Min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...