Maleic acid ≥99%

Supplier: SPECTRUM CHEMICAL MFG CORP
Warning

M1075-250GM M1075-1KG M1075-500GM M1075-5KG M1075-100GM
700005-940EA 132.39 USD
700005-940 700005-938 700005-944 700005-946 700005-936
Maleic acid ≥99%
Maleic acid

Maleic Acid is used to manufacture of artificial resins, to retard rancidity of fats and oils and in preparing the maleate salts of antihistamines and similar drugs.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₄H₄O₄
MW: 116.07 g/mol
Boiling Pt: 256 °C (1013 hPa)
Melting Pt: 130 - 131 °C
Density: 1.59 g/cm³ (20 °C)
Flash Pt: 127 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00063177
CAS Number: 110-16-7
EINECS: 203-742-5
UN: 3261
ADR: 8,III
Merck Index: 13,05726
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a3a1ea1f5bca5d5cf7fd40530f642f47db88908a&vid=1ac7f8cee2cd0433dbfb6e9271d41fe6ecc382c1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR