Magnesium nitrate hexahydrate ≥98%, crystals ACS
Supplier: SPECTRUM CHEMICAL MFG CORP
|
Warning
|
Magnesium Nitrate, Hexahydrate, Crystal, Reagent, ACS is extremely soluble in water and in ethanol. It is used in fertilizer blends in the hydroponics and greenhouse trade. It also used as a preservative, catalyst in the manufacture of petrochemicals, as a desensitizer for lithographic plates and in pyrotechnics.
As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.
Formula:
Mg(NO₃)₂·6H₂O MW: 256.41 g/mol Boiling Pt: 330 °C (1013 hPa) Melting Pt: 89 °C Density: 1.63 g/cm³ (20 °C) Flash Pt: 93.3 °C Storage Temperature: Ambient |
MDL Number:
MFCD00149779 CAS Number: 13446-18-9 EINECS: 233-826-7 UN: 1474 ADR: 5.1,III Merck Index: 13,05697 |
Specification Test Results
Assay | 98% Min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...