Magnesium chloride, anhydrous ≥98% suitable for use as excipient
Supplier: SPECTRUM CHEMICAL MFG CORP
M1018-25GM
M1018-5GM
M1018-50KGBL
700001-058EA
209.65
USD
700001-058
700001-062
700001-060
Magnesium chloride, anhydrous ≥98% suitable for use as excipient
Magnesium chloride
Magnesium Chloride, Anhydrous, is a chemical compound that is highly soluble in water and is a typical ionic halides. It has a wide range of uses, notably the production of magnesium metal, it is also used for dust control, ice control and road stabilization.
Formula:
MgCl₂ MW: 95.21 g/mol Boiling Pt: 1412 Melting Pt: 715 Density: 2.32 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011106 CAS Number: 7786-30-3 EINECS: 232-094-6 Merck Index: 13,05684 |
Specification Test Results
Assay | 98% Min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...