Light green SF yellowish

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Dihydrogen (ethyl)[4-[p-[ethyl(m-sulphonatobenzyl)amino]-p'-sulphonatobenzhydrylene]cyclohexa-2,5-dien-1-ylidene](m-sulphonatobenzyl)ammonium disodium salt, Acid Green 5, Light Green 2G, S or 2GN, CI 42095, Sodium 3-((-ethyl(-4-((4-(ethyl(3-sulphonatobenzyl)amino)phenyl)(4-sulphonatophenyl)methylene)cyclohexa-2,5-dien-1-ylidene)ammonio)methyl)benzenesulphonate, Food Green n°2, Acid Green F, Fast acid Green N, CN DcL-30

LI105-100GM LI105-10GM LI105-1KGBL LI105-5KGBL LI105-25GM
700010-088EA 542.7 USD
700010-088 700010-090 700010-092 700010-096 700010-094
Light green SF yellowish
Light green SF yellowish

Light Green SF Yellowish is a green triarylmethane dye. It is used in histology for staining collagen. It appears as brownish-red to purple powder and is soluble in cold water.

Formula: C₃₇H₃₄N₂Na₂O₉S₃
MW: 792.86 g/mol
Melting Pt: 288 - 289 °C
Storage Temperature: Ambient
MDL Number: MFCD00012121
CAS Number: 5141-20-8
EINECS: 225-906-5
Merck Index: 13,05506

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR