N-Lauroylsarcosine free acid ≥95%
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
N-Lauroyl-Sar-OH, 2-(N-Methyldodecanamido)acetic acid, N-Dodecanoylsarcosine
L3106-100GM
L3106-25GM
L3106-500GM
700007-764EA
138.4
USD
700007-764
700007-766
700007-768
N-Lauroylsarcosine free acid ≥95%
N-Lauroylsarcosine
N-Lauroylsarcosine, Free Acid is used in purifying or cleansing agents that depends on possessing both hydrophilic and hydrophobic properties.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
H₃C(CH₂)₁₀C(O)N(CH₃)CH₂COOH MW: 271.4 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00021749 CAS Number: 97-78-9 EINECS: 202-608-3 |
Specification Test Results
Assay | 95% Min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...