Methyl linoleate
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
cis,cis-9,12-Octadecadienoic acid methyl ester, 9,12-Octadecanoic acid methyl ester, 9Z,12Z-Octadecadienoic acid methyl ester, Linoleic acid methyl ester, (9Z,12Z)-Methyl octadeca-9,12-dienoate, Methyl cis,cis-9,12-octadecadienoate, 9Z,12Z-Oktadekadiensäuremethylester, 9,12 Methyl octadecaotenoate methyl linoleate, cis,cis-9,12-Oktadekadiensäuremethylester
Linoleic Acid Methyl Ester, also known as C18:2, is a polyunsaturated omega-6 fatty acid ester used in oil paints and varnishes and to exhibit the antioxidant effect of natural phenols.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
C₁₉H₃₄O₂ MW: 294.48 g/mol Boiling Pt: 340 °C (1013 hPa) Melting Pt: –35 °C Density: 0.889 Storage Temperature: Freezer |
MDL Number:
MFCD00009534 CAS Number: 112-63-0 EINECS: 203-993-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...