α-Linolenic acid, Practical grade

Supplier: Spectrum Chemical Mfg. Corp.

L1107-100GM L1107-1GM L1107-10KGBL L1107-25GM L1107-5GM
700005-450EA 2632.1 USD
700005-450 700005-454 700005-452 700005-456 700005-458
α-Linolenic acid, Practical grade
α-Linolenic acid

Linolenic Acid, Practical, a type of fatty acid, is usually found in vegetable oils.


Practical grade products supplied by Spectrum are indicative of a grade suitable for general industrial use.

Formula: C₁₈H₃₀O₂
Boiling Pt: 443 °C (1013 hPa)
Melting Pt: -9.5 °C
Density: 0.9141 g/cm³ (18 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00065720
CAS Number: 463-40-1
EINECS: 207-334-8
Merck Index: 12,05530

Order Now

Specification Test Results

Assay 70% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR