Lead(II) acetate trihydrate ≥99%, crystals ACS
Supplier: Spectrum Chemical Mfg. Corp.
|
Danger
|
Lead Acetate, Trihydrate, Crystal, Reagent, ACS is a chemical compound and is obtained by treating lead oxide with acetic acid. It is used as a reagent and comes in a crystalline form. As an ACS grade Reagent, Spectrum Chemical manufactured compounds are used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and pureness
Formula:
(H₃CCOO)₂Pb·3H₂O Boiling Pt: 280 °C (1013 hPa) Melting Pt: 75 °C Density: 2.55 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00150023 CAS Number: 6080-56-4 EINECS: 206-104-4 UN: 1616 ADR: 6.1,III Merck Index: 13,05415 |
|
Specification Test Results
Assay | 99% Min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...