Lauric acid ≥97%, Reagent Grade for synthesis

Supplier: SPECTRUM CHEMICAL MFG CORP
Warning

Synonyms: Dodecanoic acid, Dodecylic acid

L1024-2.5KG L1024-100GM L1024-500GM
700005-222EA 614.6 USD
700005-222 700005-220 700005-224
Lauric acid ≥97%, Reagent Grade for synthesis
Lauric acid

Lauric Acid, Reagent, also known as dodecanoic acid, is a medium chain fatty acid that has a vague smell of soap and is a powder. It is found naturally in human breast milk as well as cow's and goat's milk. It is generally used to produce cosmetic products but is also used in the laboratory to obtain the molar mass of substances. Its reagent grade means this is the highest quality commercially available for this chemical and that the American Chemical Society has not officially set any specifications for this material.

Formula: C₁₂H₂₄O₂
MW: 200.32 g/mol
Boiling Pt: 296
Melting Pt: 42…45 °C
Density: 0.875 g/cm³ (20 °C)
Flash Pt: >150 °C
Storage Temperature: Ambient
MDL Number: MFCD00002736
CAS Number: 143-07-7
EINECS: 205-582-1
Merck Index: 13,05400

Order Now

Specification Test Results

Assay 97% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR