Bole white (kaolin), powder USP

Supplier: Spectrum Chemical Mfg. Corp.

KA105-2.5KG KA105-500CS6 KA105-500GM KA105-12KG KA105-2.5CS4
95033-780EA 481.6 USD
95033-780 95033-782 95033-784 700000-340 95033-778
Bole white (kaolin), powder USP
Bole white (kaolin)

Melamine statement (available upon request) BSE/TSE statement (available upon request) Allergen statement (available upon request)


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: Al₂SiO₇·2H₂O
Melting Pt: 1760 °C
Density: 0.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00062311
CAS Number: 1332-58-7
EINECS: 310-194-1
REACH: exempt
Merck Index: 13,05300

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR