Isopropyl myristate ≥90% NF
Supplier: Spectrum Chemical Mfg. Corp.
Isopropyl Myristate, NF is an emollient ester of low viscosity used to reduce the greasiness of emulsions. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities
Formula:
C₁₇H₃₄O₂ Boiling Pt: 320 °C (1013 hPa) Melting Pt: 0 - 1 °C Density: 0.852 g/cm³ (20 °C) Flash Pt: 150 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008982 CAS Number: 110-27-0 EINECS: 203-751-4 Merck Index: 13,05235 |
Specification Test Results
Assay | 90% Min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...