Indigo (synthetic) for synthesis
Supplier: SPECTRUM CHEMICAL MFG CORP
|
Warning
|
Synonyms:
Pigment Blue 66, Vat Blue 1, Indigotin, 2,2'-Bis(2,3-dihydro-3-oxoindolyliden), Indigo (synthetic), 2-(1,3-Dihydro-3-oxo-2H-indol-2-ylidene)-1,2-dihydro-3H-indol-3-one
Indigo, Synthetic, or Vat Blue 1, is a synethic dye. Find a high quality inventory of synthtic indigo dye at Spectrum Chemical. Also known as Vat Blue 1, it is a vat dye and one of the oldest classes of dyes known.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
C₁₆H₁₀N₂O₂ Density: 1.48 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00005722 CAS Number: 482-89-3 EINECS: 207-586-9 Merck Index: 12,04977 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...