IGEPAL® CA-630

Supplier: SPECTRUM CHEMICAL MFG CORP
Danger

Synonyms: Octylphenyl-polyethylene glycol

I1112-100ML I1112-500ML
700010-946EA 4706.28 USD
700010-946 700010-944 700010-948
IGEPAL® CA-630
IGEPAL® CA-630

Igepal(R) CA-630 is a nonionic surfactant finding application in household and industrial controlled foam detergents, industrial metal cleaners, floor cleaners and waterless hand cleaners.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: (C₂H₄O)nC₁₄H₂₂O
Boiling Pt: ? 200 °C (1013 hPa)
Melting Pt: 6 °C
Density: 1.06 g/cm³ (20 °C)
Flash Pt: 251 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00132505
CAS Number: 9002-93-1
UN: 3082
ADR: 9,III
Merck Index: 13,06793

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR