Cetrimonium chloride ≥98%
Supplier: SPECTRUM CHEMICAL MFG CORP
|
Danger
|
Synonyms:
Cetyltrimethylammonium chloride, (1-Hexadecyl)trimethylammonium chloride, Hexadecyltrimethylammonium chloride, N-Cetyl-N,N,N-trimethylammonium chloride, N,N,N-Trimethylhexadecan-1-aminium chloride, CTAC
Hexadecyltrimethylammonium Chloride, also known as Cetrimonium bromide, is an amine based cationic quaternary surfactant used as an antiseptic agent against fungi and bacteria, and as a buffer solution for the extraction of DNA.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
C₁₉H₄₂ClN MW: 320 g/mol Melting Pt: 232…233 °C Density: 0.9 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011773 CAS Number: 112-02-7 EINECS: 203-928-6 |
Specification Test Results
Assay | 98% Min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...