8-Quinolinol hemisulfate salt, Reagent Grade

Supplier: Spectrum Chemical Mfg. Corp.
Warning

H1096-500GM H1096-100GM H1096-25GM H1096-12KG H1096-2.5KG H1096-50KGBL
700008-742EA 602.7 USD
700008-742 700008-734 700008-740 700008-736 700008-738 700008-744
8-Quinolinol hemisulfate salt, Reagent Grade
8-Quinolinol hemisulfate salt

8-Hydroxyquinoline Sulfate, Reagent is a monoprotic bidentate metal chelating agent extracted from coal tar. The resulting complexes, as well as the heterocycle itself, exhibit antiseptic, disinfectant, and pesticide properties. Its reagent grade means this is the highest quality commercially available for this chemical and that the American Chemical Society has not officially set any specifications for this material.

Formula: C₁₈H₁₄N₂O₂·H₂SO₄
Melting Pt: 177.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00013095
CAS Number: 134-31-6
EINECS: 205-137-1

Order Now

Specification Test Results

Assay 98.3% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR