D-Gluconic acid 50% in aqueous solution
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
Gluconic acid, (2R,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanoic acid
D-Gluconic Acid, 50 Percent Aqueous Solution, also known as Dextronic acid, is considered a gluconate and can be used to treat burns from hydrofluoric acid. Spectrum solutions utilize the highest quality raw materials appropriate to your product. Most raw materials meet or exceed the specifications established by the American Chemical Society. Where appropriate, many finished products are traceable to NIST Standard Reference Materials. Manufacturing, quality control testing, and packaging are all performed in Spectrum’s own facilities. Record keeping and sample retention of all produced lots ensure product consistency and complete traceability
Formula:
C₆H₁₂O₇ MW: 196.16 g/mol Density: 1.24 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00004240 CAS Number: 526-95-4 EINECS: 208-401-4 UN: 3265 ADR: 8,III |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...