7-Isopropyl-1,4-dimethylazulene ≥99%

Supplier: SPECTRUM CHEMICAL MFG CORP
Warning

Synonyms: Guaiazulene

G1143-25GM G1143-5GM
700002-358EA 189.55 USD
700002-358 700002-360
7-Isopropyl-1,4-dimethylazulene ≥99%
7-Isopropyl-1,4-dimethylazulene

Guaiazulene, also known as azulon, is a crystalline hydrocarbon and derivative of azulene.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₁₅H₁₈
MW: 198.31 g/mol
Boiling Pt: 309 °C (1013 hPa)
Melting Pt: 133 °C
Density: 0.9759 g/cm³ (18 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00003811
CAS Number: 489-84-9
EINECS: 207-701-2
Merck Index: 12,04581

Order Now

Specification Test Results

Assay 99% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR