Iron(II,III) oxide, black, powder suitable for use as excipient

Supplier: SPECTRUM CHEMICAL MFG CORP
Warning

Synonyms: Magnetite, Triiron tetraoxide, Iron(II,III) oxide

F1058-2.5KG F1058-45KGBL F1058-12KG F1058-500GM
700000-862EA 262.55 USD
700000-862 700000-864 700000-860 700000-866
Iron(II,III) oxide, black, powder suitable for use as excipient
Iron(II,III) oxide

Ferrosoferric Oxide, Black, Powder is used as the starting material for producing iron, steel and other ferro-alloys.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: Fe₃O₄
MW: 231.54 g/mol
Melting Pt: 1560
Density: 5.18 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011010
CAS Number: 1317-61-9
EINECS: 215-277-5
Merck Index: 12,04087

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR