Iron(III) oxide ≥95%, powder, Reagent Grade
Supplier: Spectrum Chemical Mfg. Corp.
|
Warning
|
Ferric Oxide, Powder, Reagent is an inorganic compound and is one of the main oxides of iron. It is used as a polishing agent (jeweler's rouge), a pigment and a photocatalyst. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.
Formula:
Fe₂O₃ Melting Pt: 1538 °C Density: 5.24 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011008 CAS Number: 1309-37-1 EINECS: 215-168-2 Merck Index: 13,04058 |
Specification Test Results
Assay | 95% Min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...