Ethyl 4-hydroxybenzoate ≥98% NF

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Nipagin A, Ethyl paraben, Ethyl p-hydroxybenzoate, Ethylparaben, Ethyl 4-hydroxybenzoate

ET117-12KG ET117-50KGBL ET117-125GM ET117-2.5KG ET117-500GM
700006-470EA 2809.55 USD
700006-470 700006-472 95033-178 95033-180 95033-182
Ethyl 4-hydroxybenzoate ≥98% NF
Ethyl 4-hydroxybenzoate

Ethylparaben, NF is used as an antimicrobial preservative. 


All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 

Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Boiling Pt: 297 °C (1013 hPa)
Melting Pt: 116…118 °C
Density: 1.078 g/cm³ (131 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002353
CAS Number: 120-47-8
EINECS: 204-399-4
Merck Index: 13,03869

Order Now

Specification Test Results

Assay 98% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR