Ethyl myristate ≥98%

Supplier: Spectrum Chemical Mfg. Corp.

E2770-500ML E2770-100ML
700005-146EA 542.7 USD
700005-146 700005-144
Ethyl myristate ≥98%
Ethyl myristate

Ethyl Myristate is a 16 carbon saturated fatty acid used as a perfumery ingredient with fresh, fruity notes.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₁₆H₃₂O₂
Density: 0.86 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008984
CAS Number: 124-06-1
EINECS: 204-675-4

Order Now

Specification Test Results

Assay 98% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR