EDTA tetrasodium salt solution

Supplier: SPECTRUM CHEMICAL MFG CORP
Danger

E1048-200LTB E1048-20LTPL E1048-1LT E1048-4LTPL
700007-306EA 1863.76 USD
700007-306 700007-308 700007-304 700007-310
EDTA tetrasodium salt solution
EDTA tetrasodium salt

Ethylenediaminetetraacetic Acid Tetrasodium Salt, Solution is typically known as EDTA and is an aminopolycarboxylic acid. It is usually used to dissolve limescale.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₁₀H₁₂N₂Na₄O₈
Melting Pt: > 300 °C
Density: 6.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00150027
CAS Number: 64-02-8
EINECS: 200-573-9
UN: 3267
ADR: 8,III
Merck Index: 12,03557
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3da8dc42f6b4faeb40c2c5e98f983e37d310fe21&vid=c925ac956246bbfd17c182e83b57c15a3b8edfa1&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR