2-Ethoxyethyl acetate ≥99%, Reagent Grade

Supplier: SPECTRUM CHEMICAL MFG CORP
Danger

Synonyms: Ethylglycol acetate

E1024-1LT E1024-200LTB E1024-20LTMT
700004-474EA 154.65 USD
700004-474 700004-476 700004-478
2-Ethoxyethyl acetate ≥99%, Reagent Grade
2-Ethoxyethyl acetate

2-Ethoxyethyl Acetate, Reagent is also known as 2-ethoxyethanol or ethyl cellosolve. It is a solvent used for industrial purposes. It is colorless and clear with barely any odor. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.

Formula: C₆H₁₂O₃
MW: 132.16 g/mol
Boiling Pt: 156 °C (1013 hPa)
Melting Pt: –62 °C
Density: 0.976 g/cm³ (20 °C)
Flash Pt: 54 °C
Storage Temperature: Ambient
MDL Number: MFCD00009251
CAS Number: 111-15-9
EINECS: 203-839-2
UN: 1172
ADR: 3,III
Merck Index: 13,03787

Order Now

Specification Test Results

Assay 99% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR