2-(2-Ethoxyethoxy)ethyl acetate ≥99%, Reagent Grade
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
Diethylene glycol monoethyl ether acetate
2-(2-Ethoxyethoxy) Ethyl Acetate, Reagent is an organic compound that is a colorless, sweet smelling liquid that is the ester of ethanol and acetic acid. It has a range of uses and is found in many products such as decaffeinating tea and coffee, cigarettes, nail polish remover, glues and is manufactured on a large scale as a solvent. In the laboratory it is used in column chromatography and extraction. Its reagent grade means this is the highest quality commercially available for this chemical and that the American Chemical Society has not officially set any specifications for this material.
Formula:
C₈H₁₆O₄ MW: 176.21 g/mol Boiling Pt: 210 °C (1013 hPa) Melting Pt: –25 °C Density: 1.0114 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00041928 CAS Number: 112-15-2 EINECS: 203-940-1 |
Specification Test Results
Assay | 99% Min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...