Diethylamine ≥98%, Reagent Grade

Supplier: Spectrum Chemical Mfg. Corp.
Danger

DI117-20LTMT DI117-200LTB DI117-4LTGL DI117-500ML
700006-744EA 3636.7 USD
700006-744 700006-742 700006-746 700006-748
Diethylamine ≥98%, Reagent Grade
Diethylamine

Diethylamine, Reagent is a secondary amine that is a flammable, volatile, strongly alkaline, colorless liquid that has a strong unpleasant smell. It is used in the production of rubber, resin, dyes and pharmaceuticals and as a corrosion inhibiter. Its Reagent grade means this is the highest quality commercially available for this chemical and that the American Chemical Society has not officially set any specifications for this material.

Formula: (C₂H₅)₂NH
Boiling Pt: 55.5 °C (1013 hPa)
Melting Pt: -50 °C
Density: 0.7048 g/cm³ (20 °C)
Flash Pt: -39 °C
Storage Temperature: Ambient
MDL Number: MFCD00009032
CAS Number: 109-89-7
EINECS: 203-716-3
UN: 1154
ADR: 3,II
Merck Index: 13,03138
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3c054fa528239af221505ef08b7c0d2503c9d1d7&vid=e1e59ef071f0c580f9eb9716f4801d314ff824c3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 98% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR