Dextran, M.W. ~ 75,000, Powder
Supplier: SPECTRUM CHEMICAL MFG CORP
DE125-100GM
DE125-1KG
DE125-25GM
700013-122EA
367.72
USD
700013-122
700013-124
700013-126
Dextran, M.W. ~ 75,000, Powder
Dextran
Dextran, 75,000, Powder is a complex branched glucan that is synthesized by certain lactic acid bacteria from sucrose. Commonly used in microsurgery to decrease the risk of vascular thrombosis, these agents are also used as effective osmotic agents and can treat hypovolemia. In the laboratory, Dextran is used to apply osmotic pressure to biological molecules and can, when applied with a fluorescent molecule, be used to create concentration gradients of diffusible molecules.
Formula:
(C₆H₁₀O₅)n Storage Temperature: Ambient |
MDL Number:
MFCD00130935 CAS Number: 9004-54-0 EINECS: 232-677-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...