N,N-Diethyl-p-phenylenediammonium sulfate ∼98% Min.
Supplier: SPECTRUM CHEMICAL MFG CORP
|
Warning
|
Synonyms:
N,N-Diethyl-p-phenylenediamine sulphate, N,N-Diethyl-1,4-phenylenediammonium sulphate, 4-Amino-N,N-diethylaniline sulphate, N1,N1-Diethylbenzene-1,4-diamine sulphate, N,N-Diethylbenzene-1,4-diammonium sulphate
D3427-100GM
D3427-500GM
700006-884EA
76.05
USD
700006-884
700006-882
700006-886
N,N-Diethyl-p-phenylenediammonium sulfate ∼98% Min.
N,N-Diethyl-p-phenylenediammonium sulphate
N,N-Diethyl-p-phenylenediamine Sulfate has applications in the laboratory including for the spectrophotometric determination of phenolic and aromatic amino groups. It has also been used as a reference probe to study rapid chlorination rate constants by a stopped-flow spectrophotometric competition kinetics method
Formula:
C₁₀H₁₈N₂O₄S MW: 262.33 g/mol Boiling Pt: 274…275 °C (1013 hPa) Melting Pt: 184…186 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012993 CAS Number: 6283-63-2 EINECS: 228-500-6 |
Specification Test Results
Assay | ∼98% Min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...