Dimethyl glutarate ≥98%
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
1,5-Pentanedioic acid dimethyl ester, Glutaric acid dimethyl ester
D2948-100GM
D2948-500GM
D2948-200LTB
700005-888EA
123.5
USD
700005-888
700005-894
700005-890
Dimethyl glutarate ≥98%
Dimethyl glutarate
Dimethyl Glutarate is used in de-icing and anti-icing applications, construction and road applications, and release and binders agents.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
C₇H₁₂O₄ MW: 160.17 g/mol Boiling Pt: 211 °C (1013 hPa) Melting Pt: –37 °C Density: 1.087 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00008468 CAS Number: 1119-40-0 EINECS: 214-277-2 Merck Index: 13,04486 |
Specification Test Results
Assay | 98% Min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...