Diethyl sebacate ≥98% suitable for use as excipient

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Sebacic acid diethyl ester, Decanedioic acid diethyl ester, Diethyl decanedioate

D2843-500ML D2843-100ML D2843-2.5LT D2843-35KGBL
700005-828EA 200.75 USD
700005-828 700005-822 700005-824 700005-826
Diethyl sebacate ≥98% suitable for use as excipient
Diethyl sebacate
Formula: C₁₄H₂₆O₄
MW: 258.36 g/mol
Boiling Pt: 312
Melting Pt: -3
Density: 0.963
Storage Temperature: Ambient
MDL Number: MFCD00009219
CAS Number: 110-40-7
EINECS: 203-764-5

Order Now

Specification Test Results

Assay 98% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR