1,12-Dodecanedioic acid ≥99%
Supplier: SPECTRUM CHEMICAL MFG CORP
|
Warning
|
Synonyms:
Dodecanedioic acid, 1,10-Decanedicarboxylic acid
D2747-100GM
D2747-500GM
700005-884EA
149.95
USD
700005-884
700005-886
1,12-Dodecanedioic acid ≥99%
1,12-Dodecanedioic acid
Dodecanedioic Acid, also known as DDDA, is a dicarboxylic acid used as an antiseptic, a corrosion inhibitor, a surfactant, and in painting materials and engineering plastics like nylon 612.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
C₁₂H₂₂O₄ MW: 230.3 g/mol Boiling Pt: 294 °C (1013 hPa) Melting Pt: 121…126 °C Density: 1.175 g/cm³ (21 °C) Flash Pt: 220 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002735 CAS Number: 693-23-2 EINECS: 211-746-3 |
Specification Test Results
Assay | 99% Min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...