2,3-Dihydroxynaphthalene ≥98%

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Naphthalene-2,3-diol, 2,3-Naphthalenediol

D2602-100GM D2602-25GM
700003-290EA 198.1 USD
700003-290 700003-292
2,3-Dihydroxynaphthalene ≥98%
2,3-Dihydroxynaphthalene
Formula: C₁₀H₈O₂
MW: 160.17 g/mol
Boiling Pt: 354 °C (1013 hPa)
Melting Pt: 151 - 152 °C
Density: 1.12 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004073
CAS Number: 92-44-4
EINECS: 202-156-7

Order Now

Specification Test Results

Assay 98% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR