Dibenzothiophene

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Dibenzo[b,d]thiophene

D1465-100GM D1465-25GM
700009-110EA 285.65 USD
700009-110 700009-112
Dibenzothiophene
Dibenzothiophene

Dibenzothiophene, also known as Diphenylene sulfide or DBT, is the organosulfur compound. It has two benzene rings which are fused to a central thiophene ring.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₁₂H₈S
MW: 184.26 g/mol
Boiling Pt: 332 °C (1013 hPa)
Melting Pt: 95…98 °C
Density: 1.1295 g/cm³ (125 °C)
Flash Pt: 170 °C
Storage Temperature: Ambient
MDL Number: MFCD00004969
CAS Number: 132-65-0
EINECS: 205-072-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR