Di-isopropanolamine

Supplier: SPECTRUM CHEMICAL MFG CORP
Warning

Synonyms: 1,1'-Iminodipropan-2-ol, Bis(2-hydroxypropyl)amine, 1,1-Iminodi-2-propanol, 1-[(2-Hydroxypropyl)amino]propan-2-ol

D1324-2.5KG D1324-500GM
700007-018EA 367.7 USD
700007-018 700007-020
Di-isopropanolamine
Di-isopropanolamine

Diisopropanolamine, also known as 1-amino-2-propanol or isopropanolamine, is an amino alcohol and used in many applications to achieve buffering, basicity, and alkalinity objectives. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₆H₁₅NO₂
MW: 133.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00004531
CAS Number: 110-97-4
EINECS: 203-820-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR