Diethylene glycol ≥97%

Supplier: SPECTRUM CHEMICAL MFG CORP
Warning

Synonyms: 2,2'-Oxybis(ethan-1-ol), Digol, 2,2'-Oxybisethanol

D1027-200LTB D1027-1LT D1027-4LT D1027-20LT
700003-442EA 2617.05 USD
700003-442 700003-440 700003-446 700003-444
Diethylene glycol ≥97%
Diethylene glycol

Diethylene Glycol is an organic compound that is colorless without any discernible odor. It is considered poisonous and is a hygroscopic liquid that tastes sweet. It is miscible in alcohol, ether, acetone, water, and ethylene glycol. It is a humectant for tobacco, cork, printing ink, and glue.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₄H₁₀O₃
MW: 106.12 g/mol
Boiling Pt: 245 °C (1013 hPa)
Melting Pt: –10 °C
Density: 1.118 g/cm³ (20 °C)
Flash Pt: 123 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00002882
CAS Number: 111-46-6
EINECS: 203-872-2
Merck Index: 13,03146
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4f023bd53ecff654f6d6d398b3ee2a1bac87d56f&vid=25d8943f9c247b4203b1e1bcba59bf77dfdd9158&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 97% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR