Cresol (mixture of isomers) CP

Supplier: SPECTRUM CHEMICAL MFG CORP
Danger

Synonyms: Hydroxy(methyl)benzene, Hydroxytoluene, Methylphenol

CR112-4LTGL CR112-500MLG
700003-218EA 473.15 USD
700003-218 700003-220
Cresol (mixture of isomers) CP
Cresol (mixture of isomers)

Cresol, CP is an organic compound also known as hydroxytoluene, a methylphenol. It can be a solid or liquid and slowly oxidized by exposure to air. Cresols are precursors or synthetic intermediates to other compounds and materials, including plastics, pharmaceuticals, and dyes. CP or Chemically Pure grade products supplied by Spectrum are indicative of a grade suitable for general industrial use. These materials may or may not have a Certificate of Analysis available.

Formula: C₇H₈O
MW: 108.14 g/mol
Boiling Pt: 200 °C (1013 hPa)
Density: 1.03 g/cm³ (20 °C)
Flash Pt: 80 °C
Storage Temperature: Ambient
MDL Number: MFCD00151099
CAS Number: 1319-77-3
EINECS: 215-293-2
UN: 2076
ADR: 6.18,II
Merck Index: 12,02645

Order Now

Specification Test Results

SPECIFIC GRAVITY @ 15.5°C 1.030 - 1.040
MOISTURE Max. 0.2%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR