Cesium chloride ≥99%, Optical Grade for spectroscopy
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
Cesium Chloride
CE117-1KG
CE117-500GM
CE117-100GM
CE117-5KGBL
CE117-25GM
700001-160EA
1542.43
USD
700001-160
700001-164
700001-158
700001-166
700001-162
Cesium chloride ≥99%, Optical Grade for spectroscopy
Cesium chloride
Cesium Chloride, Optical Grade is a colorless inorganic compound and a solid. It is used in isopynic centrifugation as well as in analytical chemistry as a reagent.
Formula:
CsCl MW: 168.36 g/mol Boiling Pt: 1303 Melting Pt: 646 Density: 3.7808 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010955 CAS Number: 7647-17-8 EINECS: 231-600-2 Merck Index: 13,02021 |
Specification Test Results
Assay | 99% Min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...