Oil of cedar wood

Supplier: SPECTRUM CHEMICAL MFG CORP
Warning

Synonyms: Cedar wood oil, Cedar Oil, Cedar Leaf Oil

CE107-500MLG CE107-100ML
700010-576EA 225.89 USD
700010-576 700010-574
Oil of cedar wood
Oil of cedar wood

Cedarwood Oil, also known as cedar leaf oil, is an essential oil used in certain medicines and perfumery. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Density: 0.98 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00146327
CAS Number: 8000-27-9

Order Now

Specification Test Results

Refractive index @20°C 1.5030-1.5100

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR