Turkey red oil

Supplier: SPECTRUM CHEMICAL MFG CORP
Warning

Synonyms: Castor oil, sulphated, Castor oil, sulphonated, Ricinus oil, sulphated

CA209-200LTB CA209-500MLG CA209-20LTPL CA209-4LTGL
700010-902EA 2923.8 USD
700010-902 700010-908 700010-904 700010-906
Turkey red oil
Turkey red oil

Castor Oil, Sulfated is an oil that is derived from the castor beans and is made by adding sulfuric acid to the oil. A colorless oil that has no taste or smell and boils at 313 C, it is used as a synthetic detergent in formulating lubricants, softeners, and dyeing assistants.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Density: 1.05 g/cm³ (20 °C)
Flash Pt: 109 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00132540
CAS Number: 8002-33-3
EINECS: 232-306-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR