Casein
Supplier: Spectrum Chemical Mfg. Corp.
CA205-2.5KG CA205-45KGBL CA205-12KG CA205-500GM
700011-216EA 413.8 USD
700011-216 700011-218 700011-214 700011-220
Casein
Proteins and Peptides
Melamine statement (available upon request)
Casein, also known as sodium caseinate, has several uses including being a component of cheese, a binder for safety matches, and as a food additive. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a certificate of analysis available.