Charcoal activated, powder USP

Supplier: Spectrum Chemical Mfg. Corp.

CA191-45KGBL CA191-12KG CA191-125GM CA191-2.5KG CA191-500GM
89078-718EA 7477 USD
89078-718 89078-716 95030-810 95030-812 95030-814
Charcoal activated, powder USP
Charcoal activated

Activated Charcoal, Powder, USP is carbon that has been treated to create low-volume pores that increase the area available for chemical reactions and adsorption. The most common pharmaceutical uses of activated charcoal is as a purification agent and antitoxin. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities

Formula: C
Melting Pt: 3550 °C
Density: 2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00133992
CAS Number: 7440-44-0
EINECS: 231-153-3
UN: 1362
ADR: 4.2,III
REACH: 01-2119488894-16
Merck Index: 13,01818

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR